missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Methanesulfonate, TRC

Product Code. 17934025
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934025 25 g 25g
17983255 1 g 1g
17913565 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17934025 Supplier Toronto Research Chemicals Supplier No. TRCI87220025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-06-7
Molecular Formula C4 H10 O3 S
Molecular Weight (g/mol) 138.19
Synonym Methanesulfonic acid 1-methylethyl ester, Methanesulfonic acid, isopropyl ester (6CI,7CI,8CI), 2-Propyl methanesulfonate, Isopropyl mesylate, Isopropyl methanesulfonate, Isopropyl methanesulphonate
IUPAC Name propan-2-yl methanesulfonate
SMILES CC(C)OS(=O)(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H10O3S/c1-4(2)7-8(3,5)6/h4H,1-3H3
Formula Weight 138.0351
Percent Purity >95%
Chemical Name or Material Isopropyl Methanesulfonate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.