missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl palmitate, tech. 85%

Product Code. 11396866
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2500 mL
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396866 2500 mL 2.5 L
11376866 250 mL 250mL
11386866 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396866 Supplier Thermo Scientific Alfa Aesar Supplier No. L04276.0F
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 142-91-6 | C19H38O2 | 298.51 g/mol

Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-91-6
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00008993
InChI Key XUGNVMKQXJXZCD-UHFFFAOYSA-N
Synonym isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit
PubChem CID 8907
ChEBI CHEBI:84262
IUPAC Name propan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Melting Point 11°C to 13°C
Density 0.852
Boiling Point 160°C (2 mmHg)
Flash Point 183°C (361°F)
Refractive Index 1.438
Quantity 2500 mL
Beilstein 1786567
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 298.51
Percent Purity 85%
Grade Technical
Chemical Name or Material Isopropyl palmitate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.