Learn More
Isopropylated phenol, phosphate (3:1), TRC
Product Code. 17993455
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993455 50 mg 50mg
17992765 5 mg 5mg
17943675 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993455 Supplier Toronto Research Chemicals Supplier No. TRCI87393550MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68937-41-7
Molecular Formula C27 H33 O4 P
Molecular Weight (g/mol) 452.52
Synonym Isopropylated Phenol, Phosphate (Mixture)
IUPAC Name Isopropylphenyl Phosphate
SMILES CC(C)C1=CC=C(C=C1)OP(=O)(OC2=CC=C(C=C2)C(C)C)OC3=CC=C(C=C3)C(C)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C27H33O4P/c1-19(2)22-7-13-25(14-8-22)29-32(28,30-26-15-9-23(10-16-26)20(3)4)31-27-17-11-24(12-18-27)21(5)6/h7-21H,1-6H3
Formula Weight 452.2116
Chemical Name or Material Tri(isopropylphenol) phosphate