missing translation for 'onlineSavingsMsg'
Learn More

Isopropyldimethylchlorosilane, 95%

Product Code. 11945841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11945841 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945841 Supplier Thermo Scientific Acros Supplier No. 313570250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3634-56-8 | C5H13ClSi | 136.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3634-56-8
Color Colorless
Density 0.8700g/mL
Boiling Point 110.0°C to 114.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C5H13ClSi
Refractive Index 1.4150 to 1.4170 (20°C, 589nm)
MDL Number MFCD00009917
Quantity 25 g
Specific Gravity 0.87
Synonym isopropyldimethylchlorosilane, dimethylisopropylchlorosilane, chloro isopropyl dimethylsilane, chloro dimethyl isopropylsilane, chlorodimethylisopropylsilane, silane,chlorodimethyl 1-methylethyl, silane, chlorodimethyl 1-methylethyl, dmipscl, acmc-1bn7i, chlorodimethyl isopropylsilane
InChI Key YCXVDEMHEKQQCI-UHFFFAOYSA-N
SMILES CC(C)[Si](C)(C)Cl
IUPAC Name chloro-dimethyl-propan-2-ylsilane
Molecular Weight (g/mol) 136.69
PubChem CID 5102883
Formula Weight 136.7
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Isopropyldimethylchlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropyldimethylchlorosilane
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.