missing translation for 'onlineSavingsMsg'
Learn More

Isopropylsulphonyl Choride, TRC

Product Code. 30391107
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391107 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391107 Supplier Toronto Research Chemicals Supplier No. TRCI8222555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropylsulphonyl Choride
CAS 10147-37-2
Molecular Formula C3H7ClO2S
Quantity 5 g
Synonym 1-Methylethylsulfonyl Chloride, 2-Propylsulfonyl Chloride, Isopropanesulfonyl Chloride, Isopropyl-2-sulfonyl Chloride, Isopropylsulfonyl Chloride, Propan-2-ylsulfonyl Chloride, Propanesulfonyl Chloride
InChI Formula InChI=1S/C3H7ClO2S/c1-3(2)7(4,5)6/h3H,1-2H3
SMILES CC(C)S(=O)(=O)Cl
IUPAC Name propane-2-sulfonyl chloride
Molecular Weight (g/mol) 142.6
Formula Weight 141.9855
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.