Learn More
Invitrogen™ ITGAV/ITGB6 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
477.00€ - 1204.00€
Specifications
| Antigen | ITGAV/ITGB6 Humanized Biosimilar |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283294
|
Invitrogen™
MA558877 |
100 μg |
477.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283928
|
Invitrogen™
MA558878 |
1 mg |
1204.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term STX-100.
Integrin subunit alpha V (ITGAV) and integrin subunit beta 6 (ITGB6) are essential components of the integrin family, which play crucial roles in cell adhesion, signaling, and tissue homeostasis. The ITGAV gene is located on chromosome 2q31, encoding the alpha-V subunit, which pairs with various beta subunits, including ITGB6, to form integrin alpha V-containing heterodimers such as alpha V beta 6. These integrins are pivotal in processes like development, immune regulation, and cancer progression. Structurally, integrins are transmembrane heterodimers comprising alpha and beta subunits that bind to extracellular matrix (ECM) proteins, mediating cell-ECM interactions. The ITGB6 gene, located on chromosome 2q24.2, encodes the beta 6 subunit, which specifically dimerizes with the alpha V subunit to form the alpha V beta 6 integrin. This heterodimer is primarily expressed in epithelial tissues and is involved in processes such as epithelial-to-mesenchymal transition (EMT), wound healing, and fibrosis. Both genes are implicated in various diseases; for example, ITGAV variants are associated with immune dysregulation and brain disorders, while ITGB6 is upregulated in cancer and plays a role in tumor progression and fibrosis.Specifications
| ITGAV/ITGB6 Humanized Biosimilar | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.