missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITLN1 Monoclonal Antibody (450520)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561408
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
ITLN1 has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. ITLN1 increases AKT phosphorylation in the absence and presence of insulin. ITLN1 may play a role in the defense system against microorganisms. ITLN1 may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. ITLN1 may be involved in iron metabolism.
Specifications
| ITLN1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8WWA0 | |
| ITLN1 | |
| E. coli-derived recombinant human Intelectin-1/Omentin, Thr19-Ser298. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 450520 | |
| Unconjugated | |
| ITLN1 | |
| calcium-dependent galactosyl-specific lectin; endothelial lectin HL-1; FLJ20022; Galactofuranose-binding lectin; hIntL; HL1; HL-1; intelectin 1; intelectin 1 (galactofuranose binding); intelectin 1 (galactofuranose binding)-like; intelectin 2; intelectin 5; intelectin a; intelectin-1; Intelectin-1a; intestinal lactoferrin receptor; INTL; Itln; ITLN1; ITLN-1; Itln1a; Itln2; Itln5; Itlna; LFR; omentin; UNQ640/PRO1270 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 55600 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction