Learn More
ITM2A Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811877
Description
This gene encodes a type II membrane protein that belongs to the ITM2 family. Studies in mouse suggest that it may be involved in osteo- and chondrogenic differentiation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| ITM2A | |
| Monoclonal | |
| Unconjugated | |
| ITM2A | |
| BRICD2A, E25A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9452 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2D5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43736 | |
| ITM2A | |
| Full length human recombit protein of human ITM2A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.