missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine Hydrochloride, TRC

Product Code. 17960835
Change view
Click to view available options
Quantity:
5 mg
100 mg
250 mg
Unit Size:
100mg
250mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960835 100 mg 100mg
17994865 5 mg 5mg
17954785 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960835 Supplier Toronto Research Chemicals Supplier No. TRCI940500100MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148849-67-6
Molecular Formula C27 H36 N2 O5 . Cl H
Molecular Weight (g/mol) 505.05
Synonym 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-, hydrochloride (1:1), 2H-3-Benzazepin-2-one, 3-[3-[[(3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl)methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-, monohydrochloride, (S)-, 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-, monohydrochloride (9CI), Bicyclo[4.2.0]octane, 2H-3-benzazepin-2-one deriv., Ivabradine hydrochloride
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
SMILES Cl.COc1cc2CCN(CCCN(C)C[C@H]3Cc4cc(OC)c(OC)cc34)C(=O)Cc2cc1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C27H36N2O5.ClH/c1-28(17-21-11-20-14-25(33-4)26(34-5)16-22(20)21)8-6-9-29-10-7-18-12-23(31-2)24(32-3)13-19(18)15-27(29)30;/h12-14,16,21H,6-11,15,17H2,1-5H3;1H/t21-;/m1./s1
Formula Weight 504.2391
Percent Purity >95%
Chemical Name or Material Ivabradine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.