Learn More
IVD Antibody (OTI1B10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71055H
Product Code. 18196983
  • 782.00€ / 0.10mL
Description
IVD Monoclonal specifically detects IVD in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IVD
Monoclonal
HRP
PBS with No Preservative
IVD
Full length recombinant protein of human IVD (NP_002612) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Primate, Monkey
IgG2b
Western Blot, Immunohistochemistry
OTI1B10
Western Blot, Immunohistochemistry
ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial
Mouse
Affinity Purified
Stem Cell Markers
3712.0
Store at 4°C in the dark.