Learn More
Avantor™ J.T. Baker™ PTFE SPE-12G column processor
Product Code. 15106056 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15106056 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15106056 Supplier Avantor™ Supplier No. 752094
  • ONLINE EXCLUSIVE PRICE
    2369.63€
    1982.12€ / Pack of 1
    Save 387.51€
    16% Off
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specially designed PTFE vacuum chambers that allow 12 samples to be run simultaneously
  • This offers the flexibility of processing SPE devices of different heights, diameters, or formats during the same experiment
  • It includes a borosilicate glass vacuum chamber, a white coloured polyamide lid including 12 Luer PTFE connectors, plugs (12×) for lid, a PE gasket, 12 Luer PP stopcocks and a PTFE sample collection rack set
  • Delivery information: Supplied with height adjustable shelves, a vacuum gauge/PTFE controller assembly, and an instruction manual
Specifications
Column Capacity (Metric) 12 samples
Type SPE-12G Column Processor
Material PTFE
Quantity 1