missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jak3 Antibody (452524) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB46991JF525
This item is not returnable.
View return policy
Description
Jak3 Monoclonal antibody specifically detects Jak3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| Jak3 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) | |
| E. coli-derived recombinant human Jak3, Gly46-Thr209, Accession # P52333 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 452524 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cell Biology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3718 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction