missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JIP1 Antibody (OTI3A12), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45033-30ul
This item is not returnable.
View return policy
Description
JIP1 Monoclonal antibody specifically detects JIP1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| JIP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI3A12 | |
| Western Blot 1:2000 | |
| C-Jun-amino-terminal kinase-interacting protein 1;IB1;JIP1;JIP-1;MAPK8IP1;PRKM8IP | |
| Full length human recombinant protein of human MAPK8IP1 (NP_005447) produced in HEK293T cell. | |
| 30 μL | |
| Apoptosis, Signal Transduction | |
| 9479 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction