missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520808
This item is not returnable.
View return policy
Description
A suggested positive control is human brain tissue lysate. PA5-20808 can be used with blocking peptide PEP-0922.
This gene encodes a nuclear protein with a JmjC domain. JmjC domain-containing proteins are predicted to function as protein hydroxylases or histone demethylases. This protein was first identified as a putative phosphatidylserine receptor involved in phagocytosis of apoptotic cells; however, subsequent studies have indicated that it does not directly function in the clearance of apoptotic cells, and questioned whether it is a true phosphatidylserine receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| JMJD6 | |
| Polyclonal | |
| Unconjugated | |
| JMJD6 | |
| 5730436I23Rik; apoptotic cell clearance receptor PtdSerR; arginine demethylase and lysine hydroxylase; bifunctional arginine demethylase and lysyl-hydroxylase JMJD6; D11Ertd195e; Histone arginine demethylase JMJD6; jmjC domain-containing protein 6; Jmjd6; jumonji domain containing 6; jumonji domain-containing protein 6; KIAA0585; Lysyl-hydroxylase JMJD6; mKIAA0585; Peptide-lysine 5-dioxygenase JMJD6; phosphatidylserine receptor; Protein PTDSR; PSR; PtdSerR; Ptdsr; PTDSR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107817, 23210, 360665 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6AYK2, Q6NYC1, Q9ERI5 | |
| JMJD6 | |
| A 15 amino acid peptide from near the amino terminus of human JMJD6. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction