missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JQ1-FITC

Product Code. 18075662
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18075662 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18075662 Supplier Tocris Bioscience™ Supplier No. 7722/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescent BET bromodomain probe; suitable for use in TR-FRET

JQ1-FITC is a fluorescent BET bromodomain probe. It binds with high affinity to BRD4(BD1) and BRD4(BD2) (KD,app values are 6.5 nM and 5.8 nM, respectively, using GST-tagged BRD4 proteins measured by CoraFluor™ TR-FRET assay; 58.7 nM for BRD4(BD1) in a fluorescence anisotropy assay). Excitation and emission maxima (λ) are 495 nm and 525 nm.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-5-(3-(1-(4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-2-oxo-7,10,13-trioxa-3-azahexadecan-16-yl)thioureido)-2-(6-hydroxy-3-oxo-3H-xanthen-9-yl)benzoic acid trifluoroacetate
Quantity 2 mg
Target Fluorescent Probes
Molecular Formula C50H50ClN7O9S2.CF3CO2H
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.