Learn More
Justrite™ Document Storage Boxes
Product Code. 11919325
Change view
Click to view available options
Dimensions (L x W x H):
1.62 x 8.75 x 11.5 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11919325 1.62 x 8.75 x 11.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11919325 Supplier Justrite™ Supplier No. 2.233.04
  • ONLINE EXCLUSIVE PRICE
    122.00€
    107.00€ / Pack of 1
    Save 15.00€
    12% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Description Medium top opening
Product Type Document Storage Box
Color Red
Material Polyethylene
Includes Label pack with 2 blank labels and labels printed SDS and instruction manual
For Use With (Application) Used to keep important papers readily available while protecting them from moisture and dirt
Certifications/Compliance OSHA
Dimensions (L x W x H) 1.62 x 8.75 x 11.5 in.
Height (English) 12.5 in.
Height (English) Interior 11.5 in.
Height (Metric) 31.8 cm
Height (Metric) Interior 29.2 cm
Length (English) 2.25 in.
Length (English) Interior 1.62 in.
Length (Metric) 5.7 cm
Length (Metric) Interior 4.1 cm
Width (English) 10.25 in.
Width (English) Interior 8.75 in.
Width (Metric) 26 cm
Width (Metric) Interior 22.2 cm