Learn More
Anritsu™ Type K Magnet Probes
Product Code. 11942139
Change view
Click to view available options
Temperature (Metric):
≤250°C
≤400°C
Style:
Miniature Round
Standard Square
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Style Unit Size
11942139 ≤250°C Miniature Round Each
11932139 ≤400°C Standard Square Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11942139 Supplier Anritsu™ Supplier No. MG11KTS1ANP
  • ONLINE EXCLUSIVE PRICE
    221.55€
    130.50€ / Pack of 1
    Save 91.05€
    41% Off
 Restricted Item
Edge
Currently Unavailable: We have no information when this product will be available.
View alternative products
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Designed with magnets to allow probes to be attached to measurement targets. Anritsu™ Type K Magnet Probes are temperature sensors designed for stationary surfaces.
  • Designed for stationary surface
  • Available with square or round footprint
  • Magnetic probe that attaches to steel surface
  • Thermocouple contact
Specifications
Accuracy ±1.3°C (100°C surface)
Analysis Time 2.0 sec.
Description Head size: 14 dia. x 24mmL
Temperature (Metric) ≤250°C
Type Thermocouple K
Material Chromel-Alumel
Style Miniature Round