missing translation for 'onlineSavingsMsg'
Learn More

K252c, TRC

Product Code. 30010045
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010045 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010045 Supplier Toronto Research Chemicals Supplier No. TRCK00051010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 85753-43-1
Molecular Formula C20 H13 N3 O
Quantity 10 mg
Synonym 6,7,12,13-Tetrahydro-5H-Indolo[2,3-a]pyrrolo[3,4-c]carbazol-5-one, Antibiotic K 252c, SD 1825, Staurosporine Aglycon, Staurosporinone
InChI Formula InChI=1S/C20H13N3O/c24-20-17-12(9-21-20)15-10-5-1-3-7-13(10)22-18(15)19-16(17)11-6-2-4-8-14(11)23-19/h1-8,22-23H,9H2,(H,21,24)
SMILES O=C1NCc2c1c3c4ccccc4[nH]c3c5[nH]c6ccccc6c25
Molecular Weight (g/mol) 311.34
Formula Weight 311.1059
Chemical Name or Material K252c
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.