Learn More
Lewis K710 Locking Safety Knife
Product Code. 17778977
Change view
Click to view available options
Color:
Black, Yellow
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17778977 Black, Yellow Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17778977 Supplier Lewis Supplier No. K710
  • ONLINE EXCLUSIVE PRICE
    609.18€
    490.72€ / Pack of 12
    Save 118.46€
    19% Off
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Lewis K710 is an industrial grade, easy-to-use, safety knife with a one-of-a-kind auto-locking safety hood that protects the user from blade exposure.
  • Once contact is lost with the cutting surface, the spring-loaded safety hood snaps shut and locks over the blade, preventing injuries common to follow-through motion.
  • The Lewis K710 features two depth settings, easy tool-free blade change and is ergonomically designed for use with either hand.
  • Attach to the CL-36 Coil Lanyard for better retention.
Specifications
Product Type Locking Safety Knife
Disposable Reusable
Sterility Non-sterile
Color Black, Yellow