Learn More
Kaempferol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30327537
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327537 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327537 Supplier Dr. Ehrenstorfer Supplier No. DREA14502000AL100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Kaempferol
CAS 520-18-3
Analyte or Component Names Kaempferol
Shipping Condition Room Temperature
Molecular Formula C15 H10 O6
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Kaempferol
InChI Formula InChI=1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)cc3
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)chromen-4-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 286.24
Quantity 1 mL
Formula Weight 286.0477 g/mol