missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525770
Additional Details : Weight : 0.01000kg
Description
KCC2 Polyclonal antibody specifically detects KCC2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
KCC2 | |
Polyclonal | |
Unconjugated | |
SLC12A5 | |
EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
57468 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9H2X9 | |
SLC12A5 | |
KLH conjugated synthetic peptide between 5-33 amino acids from the N-terminal region of human SLC12A5 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |