missing translation for 'onlineSavingsMsg'
Learn More

KCC3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA550526

Additional Details : Weight : 0.01000kg

Product Code. 15654455

  • 472.00€ / 100µL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody detects endogenous levels of total KCC3 / SLC12A6 protein.

SLC12A6 is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy.
Specifications

Specifications

KCC3
Polyclonal
Unconjugated
SLC12A6
9530023I19Rik; ACCPN; DKFZP434D2135; Electroneutral potassium-chloride cotransporter 3; furosemide-sensitive KCl cotransporter 3; gaxp; Kcc3; KCC3A; KCC3B; K-Cl cotransporter 3; potassium chloride cotransporter 3; potassium chloride cotransporter KCC3a-S3; potassium-chloride transporter-3a; potassium-chloride transporter-3b; SLC12A6; solute carrier family 12 (potassium/chloride transporter), member 6; solute carrier family 12 (potassium/chloride transporters), member 6; solute carrier family 12 member 6; solute carrier family 12, member 6
Rabbit
Antigen affinity chromatography
RUO
9990
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9UHW9
SLC12A6
Synthetic peptide corresponding to a region derived from internal residues of human Solute carrier family 12 (potassium/chloride transporters), member 6
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions