Learn More
KCC4/SLC12A7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26590-1-AP-150UL
 View more versions of this product
Product Code. 17452333
  • 410.00€ / 150µL
Description
The KCC4/SLC12A7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human KCC4/SLC12A7. This antibody recognizes human antigen. The KCC4/SLC12A7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
KCC4/SLC12A7
Polyclonal
Rabbit
Primary
Human
Western Blot, ELISA
Unconjugated
150 μL
10723