missing translation for 'onlineSavingsMsg'
Learn More

KCl 0.01D Conductivity Standards, Hach Lange™

Product Code. 10202993
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10202993 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10202993 Supplier Hach Supplier No. S51M003
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With a concentration of 1408 μS/cm ± 0.5 % at 25°C and a shelf life of 24 months from production date, each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31.

  • Features a concentration of 1408 μS/cm ± 0.5 % at 25°C
  • Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
  • According to OIML, traceable to NIST
  • Standard solution verified by Reference Materials Laboratory
  • Shelf life of 24 months from production date
  • The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
TRUSTED_SUSTAINABILITY

Specifications

Methods Standard solution verified by Reference Materials Laboratory
Analyte or Component Names KCL
Conductivity 1408μS/cm ± 0.5% at 25°C
Physical Form Liquid
Concentration 1408 μS/cm ±0.5% at 25°C
Element KCl
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.