missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK13 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41132CL1
This item is not returnable.
View return policy
Description
KCNK13 Polyclonal antibody specifically detects KCNK13 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| KCNK13 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| K2p13.1, potassium channel subfamily K member 13, potassium channel, subfamily K, member 13, tandem pore domain potassium channel THIK-1, THIK-1 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the center of human KCNK13. The immunogen is located within amino acids 150 - 200 of KCNK13. Amino Acid Squence: RRGALPQESLKDAGQ | |
| 0.1 mL | |
| Signal Transduction | |
| 56659 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction