missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21111-1-AP-150UL
This item is not returnable.
View return policy
Description
KCTD14 is a protein coding gene.Specifications
| KCTD14 | |
| Polyclonal | |
| Unconjugated | |
| KCTD14 | |
| KCTD14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233529, 65987 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BQ13 | |
| KCTD14 | |
| KCTD14 Fusion Protein Ag14265 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction