missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD9 Antibody (OTI2G8), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71968UV
This item is not returnable.
View return policy
Description
KCTD9 Monoclonal specifically detects KCTD9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| KCTD9 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KCTD9 | |
| Recombinant protein fragment corresponding to amino acids 1-223 of human KCTD9 (NP_060104) produced in E.coli. | |
| 0.1 mL | |
| 54793.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2G8 | |
| Western Blot, Immunohistochemistry | |
| BTB/POZ domain-containing protein KCTD9, FLJ20038, potassium channel tetramerisation domain containing 9 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction