missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDEL Antibody (10C3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97469SS
This item is not returnable.
View return policy
Description
KDEL Monoclonal antibody specifically detects KDEL in Human, Mouse, Rat, Avian, Chicken, Invertebrate, Mammalia, Plant, Primate, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Electron Microscopy
Specifications
| KDEL | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 50% Glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Avian, Chicken | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 10C3 | |
| Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10 - 1:500, Electron Microscopy 1:10 - 1:500 | |
| Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302 | |
| Synthetic peptide corresponding to aa 649-654 (S649EKDEL654) of rat Grp78. | |
| 0.025 mg | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| 79070 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction