Learn More
DWK Life Sciences™ KECK™ Adapter KA, complete, from hose connection glass to connection plastic
Product Code. 11781419 Shop All DWK Life Sciences Products
Click to view available options
Size:
GL 14, diameter 4.5 mm
GL 14, diameter 9.0 mm
RD 14, diameter 4.5 mm
RD 14, diameter 9.0 mm
Unit Size:
Pack of 10
Product Code. 11781419
Brand: DWK Life Sciences™ 286344203
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
KECK™ adapters prevent glass breakage on olives.
KECK™ adapters prevent glass breakage on olives and protect fingers and hands from injuries. The clamping ring, O-ring and screw cap are permanently screwed to the glass olive. The hose part can be connected quickly, securely and tightly to the threaded cap with just 1/4 turn and can also be loosened again just as quickly.
  • Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
  • permanent screw connection with the glass nozzle
  • KECK™ adapters prevent glass breakage on nozzles and protect fingers and hands from injuries
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
For Use With (Equipment) Connection of a Hose Piece with a Threaded Cap
For Use With (Application) Connection of a hose piece with a threaded cap
Size RD 14, diameter 4.5 mm