Learn More
DWK Life Sciences™ KECK™ Assembly Set
Shop All DWK Life Sciences ProductsBeskrivning
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
- With removable plastic hose connection (PBT), short and long screw (PP) and seals (VMQ, EPDM)
- Suitable for filtering flasks 100 - 20000 ml
Specifikationer
Specifikationer
| Quantity | 10 Pk. |
| Material | PBT, Polypropylene, VMQ, EPDM |
| For Use With (Application) | Filtering; Vacuum filtration; Safe working under vacuum |
| Size | 9 mm |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.