Learn More
DWK Life Sciences™ KECK™ Suction Connector, AS M8, complete with tubing parts
Product Code. 10253591 Shop All DWK Life Sciences Products
Click to view available options
Size:
4.5 mm
9 mm
Unit Size:
Pack of 10
Product Code. 10253591
Brand: DWK Life Sciences™ 286343104
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For suction or supply of liquids and gases.
The KECK™ suction nozzle with hose part is usable for suction or supply of liquids and gases from (or in) cylinders, containers, tanks, through walls with an opening of Ø 8-11 mm. Available in 2 sizes: 4.5 mm and 9 mm.
  • Suction nozzle with hose part for suction or supply of liquids and gases
  • For hoses with a diameter of Ø 8-11 mm
  • Available in 2 sizes: 4.5 mm and 9 mm.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
For Use With (Equipment) Extraction or Supply of Liquids and Gases From (or in) Cylinders, Containers, Tanks
For Use With (Application) Extraction or supply of liquids and gases from (or in) cylinders, containers, tanks
Size 9 mm