missing translation for 'onlineSavingsMsg'
Learn More

Ketones Mix 5000 μg/mL in MeOH:Water 9:1, Dr. Ehrenstorfer

Product Code. 30312806
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312806 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312806 Supplier Dr. Ehrenstorfer Supplier No. DREYS09000017MW
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butanone∣2-Hexanone∣4-Methyl-2-pentanone (MIBK)∣Acetone
CAS 78-93-3, 591-78-6, 108-10-1, 67-64-1
Analyte or Component Names 2-Butanone∣2-Hexanone∣4-Methyl-2-pentanone (MIBK)∣Acetone
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol/Water
Industry Type Food Testing
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) 2-Butanone 5000 μg/mL∣2-Hexanone 5000 μg/mL∣4-Methyl-2-pentanone (MIBK) 5000 μg/mL∣Acetone 5000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.