missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen Ethyl Ester, TRC

Product Code. 18003240
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003240 1 g 1g
18033640 5 g 5g
18013640 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18003240 Supplier Toronto Research Chemicals Supplier No. TRCK2008451G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60658-04-0
Molecular Formula C18 H18 O3
Molecular Weight (g/mol) 282.33
Synonym Ethyl 2-(3-Benzoylphenyl)propionate, Ketoprofen Ethyl Ester
IUPAC Name ethyl 2-(3-benzoylphenyl)propanoate
SMILES CCOC(=O)C(C)c1cccc(c1)C(=O)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H18O3/c1-3-21-18(20)13(2)15-10-7-11-16(12-15)17(19)14-8-5-4-6-9-14/h4-13H,3H2,1-2H3
Formula Weight 282.1256
Percent Purity >95%
Chemical Name or Material Ketoprofen Ethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.