Learn More
behr Labor-Technik™ KEX 100 F Soxhlet Extractor
Product Code. 10195450
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10195450 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 10195450 Supplier behr Labor-Technik™ Supplier No. B00217706
  • ONLINE EXCLUSIVE PRICE
    1491.00€
    872.04€ / Pack of 1
    Save 618.96€
    41% Off
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
  • Compact equipment
    • specially developed lifting tube for homogeneous extraction cycles
      • the use of extractors with a stopcock does away with the need for any additional distillation after extraction (direct redirection of solvent to the reservoir)
Specifications
Product Type Soxhlet Extractor
Capacity (Metric) 30 mL
Height (Metric) 71.5 cm
Width (Metric) 23 cm
Weight (Metric) 7.5 kg
Frequency 50/60 Hz
Voltage 230 VAC
Wattage 450 w