Learn More
behr Labor-Technik™ KEX 100 F Soxhlet Extractor System
Product Code. 10054220
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10054220 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 10054220 Supplier behr Labor-Technik™ Supplier No. B00217710
  • ONLINE EXCLUSIVE PRICE
    1761.24€
    930.91€ / Pack of 1
    Save 830.33€
    47% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Complete Kompakt single sample extraction apparatus for Soxhlet extraction. behr Labor-Technik™ KEX 100 F Soxhlet Extractor System includes a spigot.
  • With spigot
  • For 100mL fat extraction
  • One sample position
  • Complete Kompakt single sample extraction apparatus:
    • Stand
    • Heating mantle
    • Hoses
    • Glassware
      • Round-bottom flask
      • Extractor
      • Dimroth reflux condenser
  • Infinitely variable heating regulation.
  • The use of extractors with a stopcock eliminates the need for any additional distillation after extraction
  • Condensers with screwed-on tubing nipples
Specifications
Product Type Soxhlet Extractor
Capacity (Metric) 100 mL
Height (Metric) 71.5 cm
Width (Metric) 23 cm
Weight (Metric) 7.5 kg
Frequency 50/60 Hz
Voltage 230 VAC
Wattage 450 w