missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KGF Monoclonal Antibody (29568)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561166
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
Specifications
| KGF | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P21781 | |
| FGF7 | |
| E. coli-derived recombinant human KGF, Cys32-Thr194. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 29568 | |
| Unconjugated | |
| FGF7 | |
| Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction