missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
Brand: Novus Biologicals NB110-89719SS
This item is not returnable.
View return policy
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Ki67/MKI67 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.1% Sodium Azide | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| Rabbit | |
| 351 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| 4288 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot reported in scientific literature (PMID 23643677), Flow Cytometry reported in scientific literature (PMID 25733567), Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 23419702) | |
| P46013 | |
| MKI67 | |
| The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Mouse KI67/MKI67, between aminoacids 2000-2050 (2021-2036) Uniprot# E9PVX6. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction