Learn More
Ki67/MKI67 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
Brand:  Novus Biologicals NB110-89719
 View more versions of this product
Product Code. 18797943
  • 572.00€ / 0.10mL
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ki67/MKI67
Polyclonal
RUO
Western Blot, Flow Cytometry, Immunohistochemistry
The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Mouse KI67/MKI67, between aminoacids 2000-2050 (2021-2036) Uniprot# E9PVX6.