Learn More
KIAA0495 Monoclonal Antibody (OTI2A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503697S
Description
KIAA0495 Monoclonal antibody specifically detects KIAA0495 in Canine, Human, Rat samples. It is validated for Western BlotSpecifications
| KIAA0495 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9JLG9, Q9NZN9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23746, 489447, 59110 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A2 | |
| Unconjugated | |
| AIPL1 | |
| KIAA0495; PDAM; TP73 antisense RNA 1; TP73-AS1 | |
| Full length human recombinant protein of human AIPL1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.