Learn More
KIAA0513 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21478-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| KIAA0513 | |
| Polyclonal | |
| Unconjugated | |
| KIAA0513 | |
| KIAA0513 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234797, 307907, 9764 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60268, Q8R0A7 | |
| KIAA0513, RGD1304884 | |
| KIAA0513 Fusion Protein Ag15624 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |