missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0528 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21138-1-AP-150UL
This item is not returnable.
View return policy
Description
C2CD5 gene ontology annotations related to this gene include calcium-dependent phospholipid binding.Specifications
| KIAA0528 | |
| Polyclonal | |
| Unconjugated | |
| C2CD5 | |
| KIAA0528 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362461, 74741, 9847 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TPS5, Q86YS7 | |
| C2cd5 | |
| KIAA0528 Fusion Protein Ag15590 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction