Learn More
KIAA1797 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | KIAA1797 |
|---|---|
| Concentration | 850 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18038755
|
Proteintech
30140-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000656
|
Proteintech
30140-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The KIAA1797 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIAA1797. This antibody recognizes human, mouse, rat antigen. The KIAA1797 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| KIAA1797 | |
| Western Blot, ELISA | |
| Rabbit | |
| 54914 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 850 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| FLJ20375, KIAA1797 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.