missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Kidney Matched Tumor and Normal Tissue Lysate

Product Code. 18130878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18130878 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18130878 Supplier Novus Biologicals™ Supplier No. NBP247099
  • ONLINE EXCLUSIVE PRICE
    496.00€
    468.30€ / Pack of 2
    Save 27.70€
    5% Off
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Two matched 1mg/mL vials (100μg/vial) of human kidney tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.

The Kidney Matched Tumor & Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY

Specifications

Tissue Kidney
Quantity 2 Vials
Host Species Human
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.