Learn More
KIF5A Mouse anti-Human, Unconjugated, Clone: 1A7E3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67009-1-IG-150UL
 View more versions of this product
Product Code. 16839375
  • 410.00€ / 150µL
Description
The KIF5A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KIF5A. This antibody recognizes human, mouse, rat, pig antigen. The KIF5A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
KIF5A
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
1A7E3
Unconjugated
150 μL
3798