Learn More
KIF5A Mouse anti-Human, Unconjugated, Clone: 1A7E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67009-1-IG-150UL
Description
The KIF5A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KIF5A. This antibody recognizes human, mouse, rat, pig antigen. The KIF5A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| KIF5A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 1A7E3 | |
| Unconjugated | |
| 150 μL | |
| 3798 | |
| IgG1 |