Learn More
DWK Life Sciences Kimble™ KIMAX™ Erlenmeyer Flasks with Screw Caps
Product Code. 11787979
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 2
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11787979 250 mL Pack of 6
11797979 500 mL Pack of 6
11707989 1000 mL Pack of 4
11717989 2000 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11787979 Supplier DWK Life Sciences Supplier No. KC26720250
Only null left
Add to basket
Add to basket
Includes
Blue caps and pour rings
Plastic-coated for added safety
  • KimCote™ plastic coating
  • Large openings and GL 45 threaded closures for easy access and tight sealing
  • Blue caps and clear pour rings included
  • Marking spots and scales are durable white ceramic enamel
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Polypropylene
Description Reverse metric scale graduations
Diameter (English) Outer 3.18 in.
Capacity (Metric) 250 mL
Material Borosilicate Glass
Graduation Range 10 to 250 mL
Height (English) 4.92 in.
Height (Metric) 12.5 cm
Includes Clear pour rings
Writing Surface White Ceramic Enamel Marking
Closure Type Screw
Diameter (Metric) Outer 81 mm