Learn More
DWK Life Sciences Kimble™ Microfiltration Assembly Hardware
Product Code. 11863123
Change view
Click to view available options
Material:
Borosilicate Glass
Fritted Glass
Glass
Stainless Steel
Type:
Glass Support
Glass Support Base
Microfiltration Assembly
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Type unitSize
11863123 Stainless Steel Glass Support Each
11873113 Fritted Glass Glass Support Each
11878522 Glass Glass Support Base Each
11887513 Borosilicate Glass Microfiltration Assembly Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11863123 Supplier DWK Life Sciences Supplier No. 9538520001
Use Kimble™ Microfiltration Assembly Hardware when filtering large volumes of viscous or proteinaceous solutions to give the maximum flow rate.
  • Microfiltrational assembly 1000mL
  • Funnel, 4000mL
  • Flask, borosilicate glass, 47mm
  • Glass support and ground joint 90mm
  • Anodized aluminum clamp and filtration flask
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ultra-Ware 47mm Microfiltration Assembly
Material Stainless Steel
Standard Taper Joint 40/35
Type Glass Support
Length (Metric) 47 mm