Learn More
DWK Life Sciences Kimble™ Kontes™ Incubation Flasks
For metabolic studies of biological materials
Brand:  DWK Life Sciences 8823000010
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10208294
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
Ideal for measuring C14O2 yields following incubation, in vitro incubations, insulin bioassays, persulfate oxidations, and distillation of any volatile acid or base. Also suitable for use with phosphate or bicarbonate buffers when acid or alkali is introduced at the end of a reaction.
  • Available with or without sidearm
  • Flask with sidearm enables syringe needle entry without disturbing established conditions
Order Info
Stoppers and center well sold separately.
Specifications
Specifications
Rubber
Borosilicate Glass
2 in.
w/o Sidearm
3.1 cm
10 mL
For measuring C14O2 yields following incubation, in-vitro incubations, Insulin bioassays, Persulfate oxidations and distillation of any volatile Acid or base
5 cm
1.2 in.
Stopper