missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kindlin Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-523JF646
This item is not returnable.
View return policy
Description
Kindlin Polyclonal antibody specifically detects Kindlin in Human samples. It is validated for Western Blot
Specifications
| Kindlin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55612 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| C20orf42fermitin family homolog 1, chromosome 20 open reading frame 42, fermitin family homolog 1 (Drosophila), fermitin family member 1, FLJ20116, KIND1DTGCU2, kindlerin, kindlin 1, Kindlin syndrome protein, Kindlin-1, UNC112 related protein 1, UNC112A, Unc-112-related protein 1, URP1FLJ23423 | |
| Reacts with residues 643-660 (VHEYIGGYIFLSTRSKDQ) of the 77 kDa human URP1 protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction