missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kininogen 1 Mouse anti-Human, Clone: 10E8D11, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Kininogen 1 |
|---|---|
| Clone | 10E8D11 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837715
|
Proteintech
66123-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827715
|
Proteintech
66123-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| Kininogen 1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BDK, Fitzgerald factor, HMWK, Ile Ser Bradykinin Bradykinin, Kallidin I Lysyl bradykinin, kininogen 1, KNG, KNG1 | |
| KNG1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 10E8D11 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01042 | |
| 3827 | |
| Kininogen 1 Fusion Protein Ag19233 | |
| Primary | |
| -20°C | |
| KNG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title