missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ KIRREL2 293T Cell Transient Overexpression Lysate (Denatured) (T01)

Product Code. 12832263
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12832263 100 μL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12832263 Supplier Abnova™ Supplier No. CL01.0701.0100
  • ONLINE EXCLUSIVE PRICE
    75.00€
    37.20€ / 100mL
    Save 37.80€
    50% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human overexpression lysate

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description KIRREL2 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC007312.1
Molecular Weight (g/mol) Theoretical M.W. 24.2kDa
Plasmid pCMV-KIRREL2 full-length
Quality Control Testing Transient overexpression cell lysate was tested with anti-KIRREL2 antibody (Mfr. No. H00084063-B01) by Western Blot.
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • Gadolinium standard solution 10 µg/ml
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.